Thermo Scientific Chemicals

1,3-Propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 131400100
10 L, Metal drum, Each
Thermo Scientific Chemicals

1,3-Propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 131400100
10 L, Metal drum, Each
Quantity
Packaging
Catalog number: 131400100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
504-63-2
IUPAC Name
propane-1,3-diol
Molecular Formula
C3H8O2
InChI Key
YPFDHNVEDLHUCE-UHFFFAOYSA-N
SMILES
OCCCO
Color scale
=<20 APHA
GC
>=97.5 %
Infrared spectrum
Conforms
Water
=<0.2 % (K.F.)
Refractive index
1.4380 to 1.4400 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text