Thermo Scientific Chemicals

1,3-Propanedithiol, 98%, Thermo Scientific Chemicals

Catalog number: 131410500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,3-Propanedithiol, 98%, Thermo Scientific Chemicals

Catalog number: 131410500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 131410500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-80-8
IUPAC Name
propane-1,3-dithiol
Molecular Formula
C3H8S2
InChI Key
ZJLMKPKYJBQJNH-UHFFFAOYSA-N
SMILES
SCCCS
Infrared spectrum
Conforms
Refractive index
1.5380 to 1.5420 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text