2-Propanethiol, 98%, Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Propanethiol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 131440500
Price (USD)/ Each
26.40
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
26.40
Add to cart
2-Propanethiol, 98%, Thermo Scientific Chemicals
Catalog number131440500
Price (USD)/ Each
26.40
-
Add to cart
Chemical Identifiers
IUPAC Namepropane-2-thiol
Molecular FormulaC3H8S
InChI KeyKJRCEJOSASVSRA-UHFFFAOYSA-N
SMILESCC(C)S
Molecular Weight (g/mol)76.16
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4240 to 1.4270 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only