Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals
Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
250 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 131480010
Price (USD)/ Each
584.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
688.00 
Save 103.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
584.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
688.00 
Save 103.35 (15%)
Add to cart
Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals
Catalog number131480010
Price (USD)/ Each
584.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
688.00 
Save 103.35 (15%)
-
Add to cart
Chemical Identifiers
CAS106-96-7, 108-88-3
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric75 to 85 % (Product in toluene)
GC>=96.0 % (Excluding toluene)
Stabilizer0.3 % MgO
Appearance (Color)Colorless to yellow-orange
Appearance (Form)Solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only