Thermo Scientific Chemicals

Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals

Catalog number: 131480500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Propargyl bromide, 80 wt.% solution in toluene, stabilized, Thermo Scientific Chemicals

Catalog number: 131480500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 131480500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-96-7, 108-88-3
Titration Argentometric
75 to 85 % (Product in toluene)
GC
>=96.0 % (Excluding toluene)
Stabilizer
0.3 % MgO
Appearance (Color)
Colorless to yellow-orange
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text