Thermo Scientific Chemicals

Propiolic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 131501000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Propiolic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 131501000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 131501000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
471-25-0
Appearance (Color)
Clear colorless to brown
Infrared spectrum
Conforms
Titration with NaOH
97.5 to 102.5 %
Appearance (Form)
Liquid or solid
Refractive index
1.4305 to 1.4345 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text