Propylene carbonate, 99.5%, Thermo Scientific Chemicals
Propylene carbonate, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Propylene carbonate, 99.5%, Thermo Scientific Chemicals

Propylene carbonate, 99.5%, Acros Organics, Quantity: 2.5L, Packaging: Glass bottle, Boiling Point: 240.0 deg.C to 243.0 deg.C, Undesignated, Melting Point: -49.0 deg.C, Molecular Weight: 102.09, Percent Purity: 99.5%, Assay Percent Range: 99.4% min. (GC), CAS: 108-32-7, Density: 1.2040g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
500 mL
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number 131560100
Price (USD)/ Each
653.65
Online exclusive
726.00 
Save 72.35 (10%)
-
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
653.65
Online exclusive
726.00 
Save 72.35 (10%)
Add to cart
Propylene carbonate, 99.5%, Thermo Scientific Chemicals
Catalog number131560100
Price (USD)/ Each
653.65
Online exclusive
726.00 
Save 72.35 (10%)
-
Add to cart
Chemical Identifiers
CAS108-32-7
IUPAC Name4-methyl-1,3-dioxolan-2-one
Molecular FormulaC4H6O3
InChI KeyRUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILESCC1COC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<40 APHA
Infrared spectrumConforms
GC>=99.4 %
Water=<0.2 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only