Appearance of solution(10% in ethanol 96%) clear colorless to
Appearance of solutionalmost colorless
Melting point95°C to 99°C
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.