Thermo Scientific Chemicals

n-Propyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 131601000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

n-Propyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 131601000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131601000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-78-1
IUPAC Name
1-isocyanatopropane
Molecular Formula
C4H7NO
InChI Key
OQURWGJAWSLGQG-UHFFFAOYSA-N
SMILES
CCCN=C=O
Appearance (Form)
Liquid
Appearance
May develop some turbidity or precipitate
Infrared spectrum
Conforms
Appearance (Color)
Clear yellow
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text