Purine, 99%
Purine, 99%
Purine, 99%
Thermo Scientific Chemicals

Purine, 99%

CAS: 120-73-0 | C5H4N4 | 120.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 131670010
Price (TWD)
-
Request A Quote
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS120-73-0
IUPAC Name7H-purine
Molecular FormulaC5H4N4
InChI KeyKDCGOANMDULRCW-UHFFFAOYSA-N
SMILESN1C=NC2=NC=NC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to beige
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
Titration with HClO4>=98.5 %
HPLC>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470120Certificate of AnalysisMay 17, 2025131670050, 131670010
A0471268Certificate of AnalysisMay 17, 2025131670050, 131670010
A0465568Certificate of AnalysisAug 30, 2024131670050, 131670010
A0462534Certificate of AnalysisMay 13, 2024131670050, 131670010
A0458167Certificate of AnalysisDec 05, 2023131670050, 131670010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection