Purine, 99%, Thermo Scientific Chemicals
Purine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Purine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 131670050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS120-73-0
IUPAC Name7H-purine
Molecular FormulaC5H4N4
InChI KeyKDCGOANMDULRCW-UHFFFAOYSA-N
SMILESN1C=NC2=NC=NC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to beige
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
Titration with HClO4>=98.5 %
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only