2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 131711000
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
Add to cart
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
Catalog number131711000
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS98-97-5
IUPAC Namepyrazine-2-carboxylic acid
Molecular FormulaC5H4N2O2
InChI KeyNIPZZXUFJPQHNH-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point222°C to 225°C
Titration with NaOH>=98.5 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only