Thermo Scientific Chemicals

3,5-Pyrazoledicarboxylic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 131750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Pyrazoledicarboxylic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 131750050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131750050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
303180-11-2
IUPAC Name
1H-pyrazole-3,5-dicarboxylic acid hydrate
Molecular Formula
C5H6N2O5
InChI Key
GLINCONFUZIMCN-UHFFFAOYSA-N
SMILES
O.OC(=O)C1=CC(=NN1)C(O)=O
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration with NaOH
96.0 to 104.0 %
Appearance (Color)
White to off-white
Water
8.0 to 11.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text