Pyridazine, 98+%, Thermo Scientific Chemicals
Pyridazine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridazine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 131760050
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
Add to cart
Pyridazine, 98+%, Thermo Scientific Chemicals
Catalog number131760050
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
-
Add to cart
Chemical Identifiers
CAS289-80-5
IUPAC Namepyridazine
Molecular FormulaC4H4N2
InChI KeyPBMFSQRYOILNGV-UHFFFAOYSA-N
SMILESC1=CC=NN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Water=<0.5 % (K.F.)
Appearance (Color)Clear yellow-brown
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only