Thermo Scientific Chemicals

syn-2-Pyridinealdoxime, 99+%, Thermo Scientific Chemicals

Catalog number: 131791000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

syn-2-Pyridinealdoxime, 99+%, Thermo Scientific Chemicals

Catalog number: 131791000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131791000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1193-96-0
IUPAC Name
N-[(pyridin-2-yl)methylidene]hydroxylamine
Molecular Formula
C6H6N2O
InChI Key
MTFJSAGADRTKCI-UHFFFAOYSA-N
SMILES
ON=CC1=CC=CC=N1
Melting point
110°C to 113°C
Titration with HClO4
99.0 to 101.0 %
Water
=<0.2 % (K.F.)
Appearance (Color)
White to light pink
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text