2-Pyridinecarboxaldehyde, 99%, Thermo Scientific Chemicals
2-Pyridinecarboxaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Pyridinecarboxaldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 131825000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepyridine-2-carbaldehyde
Molecular FormulaC6H5NO
InChI KeyCSDSSGBPEUDDEE-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=N1
Molecular Weight (g/mol)107.11
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear yellow to yellow-brown
Appearance (Form)Liquid
Refractive index1.5343 to 1.5373 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only