Thermo Scientific Chemicals

3-Pyridinecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 131835000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Pyridinecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 131835000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131835000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
500-22-1
IUPAC Name
pyridine-3-carbaldehyde
Molecular Formula
C6H5NO
InChI Key
QJZUKDFHGGYHMC-UHFFFAOYSA-N
SMILES
O=CC1=CC=CN=C1
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.5480 to 1.5500 (20°C, 589 nm)
Appearance (Color)
Clear yellow to light brown
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text