Thermo Scientific Chemicals

4-Pyridinecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 131845000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Pyridinecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 131845000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131845000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
872-85-5
IUPAC Name
pyridine-4-carbaldehyde
Molecular Formula
C6H5NO
InChI Key
BGUWFUQJCDRPTL-UHFFFAOYSA-N
SMILES
O=CC1=CC=NC=C1
Appearance (Color)
Clear yellow-brown
Appearance (Form)
Liquid
GC
>=97.5 %
Refractive index
1.5420 to 1.5450 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text