Thermo Scientific Chemicals

2,3-Pyridinedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 131850250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

2,3-Pyridinedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 131850250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 131850250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-00-9
IUPAC Name
pyridine-2,3-dicarboxylic acid
Molecular Formula
C7H5NO4
InChI Key
GJAWHXHKYYXBSV-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CN=C1C(O)=O
Appearance (Color)
White to yellow to light beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text