Thermo Scientific Chemicals

2,4-Pyridinedicarboxylic acid hydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 131860250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Pyridinedicarboxylic acid hydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 131860250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131860250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
499-80-9
IUPAC Name
pyridine-2,4-dicarboxylic acid
Molecular Formula
C7H5NO4
InChI Key
MJIVRKPEXXHNJT-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=NC(=C1)C(O)=O
Titration with NaOH
>=99.0 % (On dry substance)
Appearance (Form)
Crystalline powder
Water
=<10 % (K.F.)
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text