2,6-Pyridinedicarboxylic Acid, 99%, Thermo Scientific Chemicals
2,6-Pyridinedicarboxylic Acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Pyridinedicarboxylic Acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 131880250
Price (USD)/ Each
34.65
Online exclusive
38.00 
Save 3.35 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
34.65
Online exclusive
38.00 
Save 3.35 (9%)
Add to cart
2,6-Pyridinedicarboxylic Acid, 99%, Thermo Scientific Chemicals
Catalog number131880250
Price (USD)/ Each
34.65
Online exclusive
38.00 
Save 3.35 (9%)
-
Add to cart
Chemical Identifiers
CAS499-83-2
IUPAC Namepyridine-2,6-dicarboxylic acid
Molecular FormulaC7H5NO4
InChI KeyWJJMNDUMQPNECX-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 %
Water=<0.3 % (K.F.)
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only