3,5-Pyridinedicarboxylic acid, 98%, Thermo Scientific Chemicals
3,5-Pyridinedicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Pyridinedicarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 131900100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS499-81-0
IUPAC Namepyridine-3,5-dicarboxylic acid
Molecular FormulaC7H5NO4
InChI KeyMPFLRYZEEAQMLQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CN=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only