Thermo Scientific Chemicals

3-Pyridinesulfonic acid, 98%, Thermo Scientific Chemicals

Catalog number: 131921000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Pyridinesulfonic acid, 98%, Thermo Scientific Chemicals

Catalog number: 131921000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 131921000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
636-73-7
IUPAC Name
pyridine-3-sulfonate
Molecular Formula
C5H4NO3S
InChI Key
DVECLMOWYVDJRM-UHFFFAOYSA-M
SMILES
[O-]S(=O)(=O)C1=CC=CN=C1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Titration with NaOH
>=97.5 %
Appearance (Color)
White to off-white
Solubility
(5 % in water) Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text