2-Pyridylacetic acid hydrochloride, 99%
2-Pyridylacetic acid hydrochloride, 99%
2-Pyridylacetic acid hydrochloride, 99%
2-Pyridylacetic acid hydrochloride, 99%
Thermo Scientific Chemicals

2-Pyridylacetic acid hydrochloride, 99%

CAS: 16179-97-8 | C7H7NO2·HCl | 173.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 131950050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16179-97-8
IUPAC Name2-(pyridin-2-yl)acetic acid hydrochloridyl
Molecular FormulaC7H7ClNO2
InChI KeyMRNWGZIWHQZCDS-UHFFFAOYSA-N
SMILES[Cl].OC(=O)CC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Titration Argentometric>=98.5 %
Appearance (Color)White to beige
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0466513Certificate of AnalysisOct 01, 2024131950250, 131950050
A0403925Certificate of AnalysisJan 02, 2024131950250, 131950050
A0374609Certificate of AnalysisDec 15, 2022131950250, 131950050
A28T041Certificate of AnalysisDec 21, 2021131950050, 131950250
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection