Thermo Scientific Chemicals

2-Pyridylacetic acid hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 131950250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Pyridylacetic acid hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 131950250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131950250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16179-97-8
IUPAC Name
2-(pyridin-2-yl)acetic acid hydrochloridyl
Molecular Formula
C7H7ClNO2
InChI Key
MRNWGZIWHQZCDS-UHFFFAOYSA-N
SMILES
[Cl].OC(=O)CC1=CC=CC=N1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Titration Argentometric
>=98.5 %
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text