Thermo Scientific Chemicals

Pyrrole-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 132060050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Pyrrole-2-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 132060050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132060050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1003-29-8
IUPAC Name
1H-pyrrole-2-carbaldehyde
Molecular Formula
C5H5NO
InChI Key
ZSKGQVFRTSEPJT-UHFFFAOYSA-N
SMILES
O=CC1=CC=CN1
Appearance (Color)
White to brown
Appearance (Form)
Low melting crystalline solid
Appearance
May darken on storage
Infrared spectrum
Conforms
Melting point
40°C to 46°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text