Thermo Scientific Chemicals

Pyrrole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 132070050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Pyrrole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 132070050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132070050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
634-97-9
IUPAC Name
1H-pyrrole-2-carboxylic acid
Molecular Formula
C5H5NO2
InChI Key
WRHZVMBBRYBTKZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CN1
Appearance (Color)
White to off-white to light brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text