Thermo Scientific Chemicals

(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 132120250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 132120250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 132120250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
98-79-3
IUPAC Name
5-oxopyrrolidine-2-carboxylic acid
Molecular Formula
C5H7NO3
InChI Key
ODHCTXKNWHHXJC-UHFFFAOYNA-N
SMILES
OC(=O)C1CCC(=O)N1
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 % (On dry substance)
Specific optical rotation
-26° to -29° (20°C, 589 nm) (c=10, 1N NaOH)
Appearance (Form)
Crystalline powder
Specific optical rotation
on dry substance

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text