Thermo Scientific Chemicals

Pyruvic acid, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 132140025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Pyruvic acid, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 132140025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 132140025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
127-17-3
IUPAC Name
2-oxopropanoic acid
Molecular Formula
C3H4O3
InChI Key
LCTONWCANYUPML-UHFFFAOYSA-N
SMILES
CC(=O)C(O)=O
Appearance (Color)
Clear colorless to yellow to amber
Titration with NaOH
97.5 to 102.5 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4260 to 1.4320 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text