Thermo Scientific Chemicals

Pyruvic acid, sodium salt, 99+%, Thermo Scientific Chemicals

Catalog number: 132150250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Pyruvic acid, sodium salt, 99+%, Thermo Scientific Chemicals

Catalog number: 132150250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 132150250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
113-24-6
IUPAC Name
sodium 2-oxopropanoate
Molecular Formula
C3H3NaO3
InChI Key
DAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES
[Na+].CC(=O)C([O-])=O
Heavy metals
=<10 ppm
Titration with HClO4
>=99.0 %
Loss on drying
=<0.5 %
Chloride (Cl)
=<20 ppm
Sulfate (SO4)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text