Pyruvic acid, sodium salt, 99+%
Pyruvic acid, sodium salt, 99+%
Pyruvic acid, sodium salt, 99+%
Pyruvic acid, sodium salt, 99+%
Thermo Scientific Chemicals

Pyruvic acid, sodium salt, 99+%

CAS: 113-24-6 | C3H3NaO3 | 110.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 132155000
Price (USD)
636.00
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
636.00
Each
Add to cart
Chemical Identifiers
CAS113-24-6
IUPAC Namesodium 2-oxopropanoate
Molecular FormulaC3H3NaO3
InChI KeyDAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES[Na+].CC(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 %
Loss on drying=<0.5 %
Chloride (Cl)=<20 ppm
Sulfate (SO4)=<20 ppm
Arsenic (As)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link