Thermo Scientific Chemicals

8-Quinolinesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 132210250
25 g, Each
Thermo Scientific Chemicals

8-Quinolinesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 132210250
25 g, Each
Quantity
Catalog number: 132210250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18704-37-5
IUPAC Name
quinoline-8-sulfonyl chloride
Molecular Formula
C9H6ClNO2S
InChI Key
JUYUYCIJACTHMK-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=C2N=CC=CC2=CC=C1
Appearance (Form)
Crystalline powder
Appearance (Color)
White to beige
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 %
Melting point
126°C to 130°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text