Resorcinol, 98%
Resorcinol, 98%
Resorcinol, 98%
Thermo Scientific Chemicals

Resorcinol, 98%

CAS: 108-46-3 | C6H6O2 | 110.11 g/mol
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 132290050
Price (JPY)
-
Request A Quote
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS108-46-3
IUPAC Namebenzene-1,3-diol
Molecular FormulaC6H6O2
InChI KeyGHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILESOC1=CC(O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or granules or flakes
Melting point108°C to 112°C
Appearance (Color)White to yellow to beige to pink
Infrared spectrumConforms
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473193Certificate of AnalysisJun 04, 2025132292500, 132290500, 132290050, 132290010
A0464898Certificate of AnalysisAug 02, 2024132292500, 132290500, 132290050, 132290010
A0460294Certificate of AnalysisFeb 16, 2024132292500, 132290500, 132290050, 132290010
A0393534Certificate of AnalysisDec 15, 2022132292500, 132290500, 132290050, 132290010
A0412933Certificate of AnalysisDec 13, 2022132292500, 132290500, 132290050, 132290010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection