Rhodamine B
Thermo Scientific Chemicals

Rhodamine B

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic drum
Catalog number 132310025
Price (USD)/ Each
564.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
664.00 
Save 99.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
564.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
664.00 
Save 99.35 (15%)
Add to cart
Rhodamine B
Catalog number132310025
Price (USD)/ Each
564.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
664.00 
Save 99.35 (15%)
-
Add to cart
Chemical Identifiers
CAS81-88-9
IUPAC Name9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC28H31ClN2O3
InChI KeyPYWVYCXTNDRMGF-UHFFFAOYSA-N
SMILES[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Infrared spectrumConforms
Absorption Ratio P-15/P+151.1 to 1.6
Loss on drying=<10 % (105°C)
Specific absorbance A (1%/1cm)>=1700 (at lambda max.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only