Chemical Identifiers
CAS83-88-5
IUPAC Name7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
Molecular FormulaC17H20N4O6
InChI KeyAUNGANRZJHBGPY-QTZZOOGMNA-N
SMILESCC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1
View more