Thermo Scientific Chemicals

Riboflavin, 98%

Catalog number: 132351000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Riboflavin, 98%

Catalog number: 132351000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132351000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-88-5
IUPAC Name
7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
Molecular Formula
C17H20N4O6
InChI Key
AUNGANRZJHBGPY-QTZZOOGMNA-N
SMILES
CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1
Appearance (Form)
Powder
UV spectrophotometry
(A (1%/1 cm) 328, on dried substance)
Loss on drying
=<1.5 % (105°C)
Ratio of Absorbances
0.31 to 0.33 (A373/A267)
Additional info
lumiflavine: passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text