Thermo Scientific Chemicals

D(-)-Ribose, 99+%

Catalog number: 132360050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

D(-)-Ribose, 99+%

Catalog number: 132360050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132360050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-69-1
IUPAC Name
(3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
Molecular Formula
C5H10O5
InChI Key
HMFHBZSHGGEWLO-ZHZWXSSZNA-N
SMILES
OC[C@H]1OC(O)[C@H](O)[C@@H]1O
Loss on drying
=<1 % (60°C, 3 h)
Specific optical rotation
-18° to -22° (20°C, 589 nm) (c=2, water)
Appearance (Color)
White to light beige
Appearance (Form)
Powder or crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text