Thermo Scientific Chemicals

Rubrene, 99%, Thermo Scientific Chemicals

Catalog number: 132380010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Rubrene, 99%, Thermo Scientific Chemicals

Catalog number: 132380010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132380010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
517-51-1
IUPAC Name
5,6,11,12-tetraphenyltetracene
Molecular Formula
C42H28
InChI Key
YYMBJDOZVAITBP-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=C2C(C3=CC=CC=C3)=C3C=CC=CC3=C(C3=CC=CC=C3)C2=C(C2=CC=CC=C2)C2=CC=CC=C12
Infrared spectrum
Conforms
HPLC
>=98.5 %
Appearance (Color)
Red
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text