Thermo Scientific Chemicals

Salicylaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 132600010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Salicylaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 132600010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 132600010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-02-8
IUPAC Name
2-hydroxybenzaldehyde
Molecular Formula
C7H6O2
InChI Key
SMQUZDBALVYZAC-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1C=O
Phenol
=<1.0 %
Infrared spectrum
Conforms
GC
>=98.5 %
Color scale
=<200 APHA
Refractive index
1.5715 to 1.5745 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text