Salicylaldehyde, 99%, Thermo Scientific Chemicals
Salicylaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Salicylaldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 132602500
Price (USD)/ Each
50.65
Online exclusive
59.40 
Save 8.75 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
50.65
Online exclusive
59.40 
Save 8.75 (15%)
Add to cart
Salicylaldehyde, 99%, Thermo Scientific Chemicals
Catalog number132602500
Price (USD)/ Each
50.65
Online exclusive
59.40 
Save 8.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeySMQUZDBALVYZAC-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C=O
Molecular Weight (g/mol)122.12
View more
SpecificationsSpecification SheetSpecification Sheet
Phenol=<1.0 %
Infrared spectrumConforms
GC>=98.5 %
Color scale=<200 APHA
Refractive index1.5715 to 1.5745 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only