Sebacic Acid, 98%, Thermo Scientific Chemicals
Sebacic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sebacic Acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 132630010
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
Add to cart
Sebacic Acid, 98%, Thermo Scientific Chemicals
Catalog number132630010
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS111-20-6
IUPAC Namedecanedioic acid
Molecular FormulaC10H18O4
InChI KeyCXMXRPHRNRROMY-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Ash=<0.06 %
Water=<0.3 %
Appearance (Form)Powder or granules
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only