Thermo Scientific Chemicals

L-Serine, 99%

Catalog number: 132661000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-Serine, 99%

Catalog number: 132661000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 132661000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-45-1
IUPAC Name
2-amino-3-hydroxypropanoic acid
Molecular Formula
C3H7NO3
InChI Key
MTCFGRXMJLQNBG-UHFFFAOYNA-N
SMILES
NC(CO)C(O)=O
Other amino acids
=<0.5 % (TLC)
Specific optical rotation
+14.4° to +16.0° (20°C, 589 nm) (c=10, 2 N HCl)
Arsenic (As)
=<1 ppm
pH
5.0 to 6.5 (2.5 % at 25°C)
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text