Chemical Identifiers
CAS61-47-2
IUPAC Name2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one 3-(2-aminoethyl)-1H-indol-5-ol sulfuric acid hydrate
Molecular FormulaC14H23N5O7S
InChI KeyBKCXVJIGPVULPX-UHFFFAOYSA-N
SMILESO.OS(O)(=O)=O.CN1CC(=O)N=C1N.NCCC1=CNC2=CC=C(O)C=C12
View more