Thermo Scientific Chemicals

Sesamol, 98%, Thermo Scientific Chemicals

Catalog number: 132690050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Sesamol, 98%, Thermo Scientific Chemicals

Catalog number: 132690050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132690050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
533-31-3
IUPAC Name
2H-1,3-benzodioxol-5-ol
Molecular Formula
C7H6O3
InChI Key
LUSZGTFNYDARNI-UHFFFAOYSA-N
SMILES
OC1=CC=C2OCOC2=C1
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder
Water
=<0.2 % (K.F.)
Appearance (Color)
Off-white to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text