Thermo Scientific Chemicals

Shikimic Acid, 98%

Catalog number: 132700050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Shikimic Acid, 98%

Catalog number: 132700050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132700050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
138-59-0
IUPAC Name
3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid
Molecular Formula
C7H10O5
InChI Key
JXOHGGNKMLTUBP-UHFFFAOYNA-N
SMILES
OC1CC(=CC(O)C1O)C(O)=O
Infrared spectrum
Conforms
Specific optical rotation
-170° to -185° (20°C, 589 nm) (C=2, H2O)
Appearance (Color)
White to light beige
Melting point
183°C to 191°C
Titration with NaOH
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text