Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

D-Sorbitol, 97%

Catalog number: 132730025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

D-Sorbitol, 97%

Catalog number: 132730025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 132730025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-70-4
IUPAC Name
hexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H14O6
InChI Key
FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)C(O)CO
HPLC
>=96.0 %
Sulfated ash
=<0.1 %
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Specific optical rotation
+4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text