Thermo Scientific Chemicals

Spermine, 97%, Thermo Scientific Chemicals

Catalog number: 132750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Spermine, 97%, Thermo Scientific Chemicals

Catalog number: 132750050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 132750050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-44-3
IUPAC Name
(3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine
Molecular Formula
C10H26N4
InChI Key
PFNFFQXMRSDOHW-UHFFFAOYSA-N
SMILES
NCCCNCCCCNCCCN
Appearance (Form)
Crystalline powder or low melting mass
Titration with HClO4
>=96.0 % (On dry substance)
Water
=<3 % (K.F.)
Appearance (Color)
White to light yellow
Melting point
26°C to 30°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text