Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 132790010
Price (USD)/ Each
47.65
Online exclusive
55.40 
Save 7.75 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
55.40 
Save 7.75 (14%)
Add to cart
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Catalog number132790010
Price (USD)/ Each
47.65
Online exclusive
55.40 
Save 7.75 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Nameethenylbenzene
Molecular FormulaC8H8
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1
Molecular Weight (g/mol)104.15
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Peroxides=<0.01 % (as H2O2)
Appearance (Form)Clear liquid
Infrared spectrumConforms
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only