Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
10 mL
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number 132790250
Price (USD)/ Each
486.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
572.00 
Save 85.35 (15%)
-
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (USD)/ Each
486.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
572.00 
Save 85.35 (15%)
Add to cart
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
Catalog number132790250
Price (USD)/ Each
486.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
572.00 
Save 85.35 (15%)
-
Add to cart
Chemical Identifiers
CAS100-42-5
IUPAC Nameethenylbenzene
Molecular FormulaC8H8
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Peroxides=<0.01 % (as H2O2)
Appearance (Form)Clear liquid
Infrared spectrumConforms
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only