Sulfur trioxide pyridine complex, technical, 48-50% active SO3, Thermo Scientific Chemicals
Sulfur trioxide pyridine complex, technical, 48-50% active SO3, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfur trioxide pyridine complex, technical, 48-50% active SO3, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 132871000
Price (USD)/ Each
86.65
Online exclusive
96.00 
Save 9.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
96.00 
Save 9.35 (10%)
Add to cart
Sulfur trioxide pyridine complex, technical, 48-50% active SO3, Thermo Scientific Chemicals
Catalog number132871000
Price (USD)/ Each
86.65
Online exclusive
96.00 
Save 9.35 (10%)
-
Add to cart
Chemical Identifiers
CAS26412-87-3
IUPAC Namepyridine; sulfonylideneoxidane
Molecular FormulaC5H5NO3S
InChI KeyUDYFLDICVHJSOY-UHFFFAOYSA-N
SMILESO=S(=O)=O.C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay95 to 100 % (Sulfur trioxide pyridine)
Appearance (Color)White to beige
Infrared spectrumConforms
Assay48 to 50 % (Active SO3)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only